Biphenyl-3,3'-dicarbonsäure

Lieferant: COMBI-BLOCKS

CA-4862-1G CA-4862-250M
COBBCA-4862-1GEA 310 EUR
COBBCA-4862-1G COBBCA-4862-250M
Biphenyl-3,3'-dicarbonsäure
Biphenyl-3,3'-dicarbonsäure
Formel: C₁₄H₁₀O₄
Molecular Weight: 242.23 g/mol
MDL: MFCD00845987
CAS-Nummer: 612-87-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR