Biphenyl-3,4'-dicarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: [1,1'-Biphenyl]-3,4'-dicarboxylic acid

CA-4861-1G CA-4861-250M
COBBCA-4861-1GEA 310 EUR
COBBCA-4861-1G COBBCA-4861-250M
Biphenyl-3,4'-dicarbonsäure
Biphenyl-3,4'-dicarbonsäure
Formel: C₁₄H₁₀O₄
Molecular Weight: 242.23 g/mol
MDL: MFCD00728324
CAS-Nummer: 92152-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR