Biphenyl-4,4'-dicarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4,4'-Biphenyldicarboxylic acid, [1,1'-Biphenyl]-4,4'-dicarboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

225266-25G 225266-5G
SIAL225266-25GEA 476 EUR
SIAL225266-25G SIAL225266-5G
Biphenyl-4,4'-dicarbonsäure, Sigma-Aldrich®
Biphenyl-4,4'-dicarbonsäure
Formel: C₁₄H₁₀O₄
Molecular Weight: 242.23 g/mol
MDL: MFCD00002554
CAS-Nummer: 787-70-2
EINECS: 212-328-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR