2'-Fluorbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4824-5G CA-4824-1G
COBBCA-4824-5GEA 451 EUR
COBBCA-4824-5G COBBCA-4824-1G
2'-Fluorbiphenyl-3-carbonsäure
2'-Fluorbiphenyl-3-carbonsäure
Formel: C₁₃H₉FO₂
Molecular Weight: 216.21
MDL: MFCD03424610
CAS-Nummer: 103978-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR