3'-Fluorbiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4822-5G CA-4822-1G
COBBCA-4822-5GEA 130 EUR
COBBCA-4822-5G COBBCA-4822-1G
3'-Fluorbiphenyl-3-carbonsäure
3'-Fluorbiphenyl-3-carbonsäure
Formel: C₁₃H₉FO₂
Molecular Weight: 216.21
MDL: MFCD03424608
CAS-Nummer: 168619-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR