4'-Fluorbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

014260-100MG 014260-1G
FLUO014260-100MGEA 0 EUR
FLUO014260-100MG FLUO014260-1G
4'-Fluorbiphenyl-3-carbonsäure
4'-Fluorbiphenyl-3-carbonsäure
Formel: C₁₃H₉FO₂
Molecular Weight: 216.21
MDL: MFCD00452724
CAS-Nummer: 10540-39-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR