2'-Fluorbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2'-Fluoro-[1,1'-biphenyl]-4-carboxylic acid

014264-100MG 014264-1G
FLUO014264-100MGEA 90.9 EUR
FLUO014264-100MG FLUO014264-1G
2'-Fluorbiphenyl-4-carbonsäure
2'-Fluorbiphenyl-4-carbonsäure
Formel: C₁₃H₉FO₂
Molecular Weight: 216,21 g/mol
MDL: MFCD00278732
CAS-Nummer: 365-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR