3'-Fluorbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Fluoro-[1,1'-biphenyl]-4-carboxylic acid

014261-100MG 014261-1G
FLUO014261-100MGEA 0 EUR
FLUO014261-100MG FLUO014261-1G
3'-Fluorbiphenyl-4-carbonsäure
3'-Fluorbiphenyl-4-carbonsäure
Formel: C₁₃H₉FO₂
Molecular Weight: 216.21
MDL: MFCD07779362
CAS-Nummer: 137045-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR