2'-Fluor-5'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4939-5G CA-4939-1G
COBBCA-4939-5GEA 2510 EUR
COBBCA-4939-5G COBBCA-4939-1G
2'-Fluor-5'-methoxybiphenyl-3-carbonsäure
2'-Fluor-5'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁FO₃
Molecular Weight: 246.24
MDL: MFCD15143531
CAS-Nummer: 1215205-32-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR