3'-Fluor-4'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4951-1G CA-4951-5G
COBBCA-4951-1GEA 204 EUR
COBBCA-4951-1G COBBCA-4951-5G
3'-Fluor-4'-methoxybiphenyl-3-carbonsäure
3'-Fluor-4'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁FO₃
Molecular Weight: 246.24
MDL: MFCD06858733
CAS-Nummer: 885964-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR