3'-Fluor-5'-methoxybiphenyl-3-carbonsäure
Lieferant: COMBI-BLOCKS
CA-4938-5G
CA-4938-1G
CA-4938-250M
COBBCA-4938-5GEA
884
EUR
COBBCA-4938-5G
COBBCA-4938-1G
COBBCA-4938-250M
3'-Fluor-5'-methoxybiphenyl-3-carbonsäure
3'-Fluor-5'-methoxybiphenyl-3-carbonsäure
Formel:
C₁₄H₁₁FO₃ Molecular Weight: 246.24 |
MDL:
MFCD15143530 CAS-Nummer: 1215205-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...