4'-Fluor-3'-methoxybiphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

CA-4937-1G CA-4937-5G CA-4937-250M
COBBCA-4937-1GEA 303 EUR
COBBCA-4937-1G COBBCA-4937-5G COBBCA-4937-250M
4'-Fluor-3'-methoxybiphenyl-3-carbonsäure
4'-Fluor-3'-methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₁FO₃
Molecular Weight: 246.24
MDL: MFCD15143529
CAS-Nummer: 1215206-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR