5-Phenyl-o-anisidin
Lieferant: FLUOROCHEM
Synonyme:
2-Amino-4-phenylanisole, 2-Methoxy-5-phenylaniline, 5-Phenyl-o-anisidine, 3-Amino-4-methoxybiphenyl
018955-1G
018955-5G
FLUO018955-1GEA
0
EUR
FLUO018955-1G
FLUO018955-5G
5-Phenyl-o-anisidin
5-Phenyl-o-anisidin
Formel:
C₁₃H₁₃NO Molecular Weight: 199,25 g/mol |
MDL:
MFCD00007790 CAS-Nummer: 39811-17-1 EINECS: 254-639-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...