Diethylcyclopropan-1,1-dicarboxylat

Lieferant: CHEMPUR

NRB03020.250 NRB03020.1
CHMPNRB03020.250EA 0 EUR
CHMPNRB03020.250 MAYBNRB03020.1
Diethylcyclopropan-1,1-dicarboxylat
Diethylcyclopropan-1,1-dicarboxylat
Formel: C₉H₁₄O₄
MDL: MFCD00001283
CAS-Nummer: 1559-02-0
EINECS: 216-321-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR