Dimethylcyclopropan-1,1-dicarboxylat

Lieferant: FLUOROCHEM

Synonyme: Dimethyl 1,1-cyclopropanedicarboxylate, 1,1-Cyclopropanedicarboxylic acid dimethyl ester

013935-25G 013935-100G
FLUO013935-25GEA 112 EUR
FLUO013935-25G FLUO013935-100G
Dimethylcyclopropan-1,1-dicarboxylat
Dimethylcyclopropan-1,1-dicarboxylat
Formel: C₇H₁₀O₄
Molecular Weight: 158.15 g/mol
MDL: MFCD00192079
CAS-Nummer: 6914-71-2
EINECS: 414-240-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR