2-Naphthalinthiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Thionaphthol

MQ100

270849-25G 270849-5G
SIAL270849-25GEA 173 EUR
SIAL270849-25G SIAL270849-5G
2-Naphthalinthiol, Sigma-Aldrich®
2-Naphthalinthiol
Formel: C₁₀H₈S
Molecular Weight: 160.24 g/mol
MDL: MFCD00004086
CAS-Nummer: 91-60-1
EINECS: 202-082-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR