3-Iodbenzaldehyd ≥98%
Lieferant: Thermo Scientific
L17447.03
L17447.06
L17447.03EA
88.4
EUR
L17447.03
L17447.06
3-Iodbenzaldehyd ≥98%
3-Iodobenzaldehyde
Purity: 98%
CAS No.: 696-41-3
Molecular Formula: C7H5IO
Formula Weight: 232.02
Storage Temperature: 2°C to 8°C
Melting Point: 55-57°
Boiling Point: 124-125°/13mm
Beilstein Reference: 1854653
MDL No.: MFCD00039573
Formel:
C₇H₅IO Molecular Weight: 232,02 g/mol Siedepunkt: 124…125 °C (13 mmHg) Schmelzpunkt: 55…57 °C |
MDL:
MFCD00039573 CAS-Nummer: 696-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...