Ethylisothiocyanatformiat
Lieferant: FLUOROCHEM
Synonyme: Ethoxycarbonyl isothiocyanate
002560-5G 002560-25G
FLUO002560-5GEA 72.9 EUR
FLUO002560-5G FLUO002560-25G
Ethylisothiocyanatformiat
Ethylisothiocyanatformiat
Formel: C₄H₅NO₂S
Molecular Weight: 131,16 g/mol
MDL: MFCD00004814
CAS-Nummer: 16182-04-0
EINECS: 240-318-9