3-(Trimethylsilyl)-2-propin-1-ol

Lieferant: CHEMPUR SILANE

Synonyme: 3-(Trimethylsilyl)propargyl alcohol

SILS20100.1 SILS20100.5 SILS20100.25
SILNSILS20100.1EA 0 EUR
SILNSILS20100.1 SILNSILS20100.5 SILNSILS20100.25
3-(Trimethylsilyl)-2-propin-1-ol
3-(Trimethylsilyl)-2-propin-1-ol
Formel: C₆H₁₂OSi
Molecular Weight: 128.25 g/mol
MDL: MFCD00002913
CAS-Nummer: 5272-36-6
EINECS: 226-094-5
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR