2-Cyanphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Cyanobenzeneboronic acid pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 2-Cyanophenylboronic acid, pinacol ester

PN-2471-1G PN-2471-5G
COBBPN-2471-1GEA 46.8 EUR
COBBPN-2471-1G COBBPN-2471-5G
2-Cyanphenylboronsäurepinakolester
2-Cyanphenylboronsäurepinakolester
Formel: C₁₃H₁₆BNO₂
Molecular Weight: 229,09 g/mol
MDL: MFCD04038747
CAS-Nummer: 214360-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR