2-Cyanphenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Cyanobenzeneboronic acid pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 2-Cyanophenylboronic acid, pinacol ester
M-Clarity™ Qualitätsstufe = MQ100
683884-1G
683884-5G
SIAL683884-1GEA
52.8
EUR
SIAL683884-1G
SIAL683884-5G
2-Cyanphenylboronsäurepinakolester, Sigma-Aldrich®
2-Cyanphenylboronsäurepinakolester
Formel:
C₁₃H₁₆BNO₂ Molecular Weight: 229,09 g/mol |
MDL:
MFCD04038747 CAS-Nummer: 214360-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...