3-Cyanphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 3-Cyanobenzeneboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

578401-1G 578401-5G
SIAL578401-1GEA 49.3 EUR
SIAL578401-1G SIAL578401-5G
3-Cyanphenylboronsäurepinakolester, Sigma-Aldrich®
3-Cyanphenylboronsäurepinakolester
Formel: C₁₃H₁₆BNO₂
Molecular Weight: 229,09 g/mol
MDL: MFCD03789262
CAS-Nummer: 214360-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR