3-Cyanphenylboronsäurepinakolester

Lieferant: FLUOROCHEM

010830-5G 010830-1G
FLUO010830-5GEA 67.1 EUR
FLUO010830-5G FLUO010830-1G
3-Cyanphenylboronsäurepinakolester
3-Cyanphenylboronsäurepinakolester
Formel: C₁₃H₁₆BNO₂
MDL: MFCD03789262
CAS-Nummer: 214360-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR