3-Cyan-5-methoxyphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-Cyano-5-methoxyphenylboronic acid, pinacol ester, 3-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
PN-5504-1G
PN-5504-250M
COBBPN-5504-1GEA
619
EUR
COBBPN-5504-1G
COBBPN-5504-250M
3-Cyan-5-methoxyphenylboronsäurepinakolester
3-Cyan-5-methoxyphenylboronsäurepinakolester
Formel:
C₁₄H₁₈BNO₃ Molecular Weight: 259,11 g/mol |
MDL:
MFCD11855976 CAS-Nummer: 1035266-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...