2-(N,N-Dimethylamino)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N,N-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

PN-8867-1G PN-8867-5G
COBBPN-8867-1GEA 0 EUR
COBBPN-8867-1G COBBPN-8867-5G
2-(N,N-Dimethylamino)phenylboronsäurepinakolester
2-(N,N-Dimethylamino)phenylboronsäurepinakolester
Formel: C₁₄H₂₂BNO₂
MDL: MFCD05864304
CAS-Nummer: 832114-08-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR