2-Aminophenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester

010824-2G 010824-5G FL10824.20 010824-25G
FLUO010824-2GEA 0 EUR
FLUO010824-2G FLUO010824-5G CHMPFL10824.20 FLUO010824-25G
2-Aminophenylboronsäurepinakolester
2-Aminophenylboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
Molecular Weight: 219.09 g/mol
MDL: MFCD02179448
CAS-Nummer: 191171-55-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR