2-Aminophenylboronsäurepinakolester 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester
367560010
367560050
ACRO367560010EA
79.8
EUR
ACRO367560010
ACRO367560050
2-Aminophenylboronsäurepinakolester 97%
2-Aminophenylboronsäurepinakolester
Formel:
C₁₂H₁₈BNO₂ Molecular Weight: 219,09 g/mol Schmelzpunkt: 64…70 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179448 CAS-Nummer: 191171-55-8 |
Spezifikation Testergebnisse
Appearance | Cream Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 64 to 70 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...