2-Aminophenylboronsäurepinakolester ≥98%

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester

BB-2121-5G BB-2121-1G
COBBBB-2121-5GEA 127 EUR
COBBBB-2121-5G COBBBB-2121-1G
2-Aminophenylboronsäurepinakolester ≥98%
2-Aminophenylboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
Molecular Weight: 219,09 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02179448
CAS-Nummer: 191171-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR