3-Aminophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester

PN-2141-25G PN-2141-5G PN-2141-1G
COBBPN-2141-25GEA 308 EUR
COBBPN-2141-25G COBBPN-2141-5G COBBPN-2141-1G
3-Aminophenylboronsäurepinakolester
3-Aminophenylboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
Molecular Weight: 219,09 g/mol
MDL: MFCD03453668
CAS-Nummer: 210907-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR