4-Aminophenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

518751-1G 518751-5G
SIAL518751-1GEA 42.8 EUR
SIAL518751-1G SIAL518751-5G
4-Aminophenylboronsäurepinakolester, Sigma-Aldrich®
4-Aminophenylboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
Molecular Weight: 219,09 g/mol
MDL: MFCD02093721
CAS-Nummer: 214360-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR