4-Aminophenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester
010826-5G
FL10826.20
010826-25G
010826-1G
FLUO010826-5GEA
57.8
EUR
FLUO010826-5G
CHMPFL10826.20
FLUO010826-25G
FLUO010826-1G
4-Aminophenylboronsäurepinakolester
4-Aminophenylboronsäurepinakolester
Formel:
C₁₂H₁₈BNO₂ Molecular Weight: 219,09 g/mol |
MDL:
MFCD02093721 CAS-Nummer: 214360-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...