4-Aminophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester

PN-2135-5G PN-2135-25G PN-2135-1G
COBBPN-2135-5GEA 50.9 EUR
COBBPN-2135-5G COBBPN-2135-25G COBBPN-2135-1G
4-Aminophenylboronsäurepinakolester
4-Aminophenylboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
Molecular Weight: 219,09 g/mol
MDL: MFCD02093721
CAS-Nummer: 214360-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR