4-Aminophenylboronsäurepinakolester 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester

366380010 366380050
ACRO366380010EA 76.6 EUR
ACRO366380010 ACRO366380050
4-Aminophenylboronsäurepinakolester 97%
4-Aminophenylboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
Molecular Weight: 219,09 g/mol
Schmelzpunkt: 165…170 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02093721
CAS-Nummer: 214360-73-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 165 to 170 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR