2-Aminophenylboronsäurepinakolester Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Aminobenzeneboronic acid pinacol ester hydrochloride

BB-2122-5G BB-2122-1G
COBBBB-2122-5GEA 0 EUR
COBBBB-2122-5G COBBBB-2122-1G
2-Aminophenylboronsäurepinakolester Hydrochlorid
2-Aminophenylboronsäurepinakolester Hydrochlorid
Formel: C₁₂H₁₉BClNO₂
MDL: MFCD02179449
CAS-Nummer: 393877-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR