3-Aminophenylboronsäurepinakolester Hydrochlorid

Lieferant: COMBI-BLOCKS

PN-2229-5G PN-2229-1G
COBBPN-2229-5GEA 460 EUR
COBBPN-2229-5G COBBPN-2229-1G
3-Aminophenylboronsäurepinakolester Hydrochlorid
3-Aminophenylboronsäurepinakolester Hydrochlorid
Formel: C₁₂H₁₉BClNO₂
Molecular Weight: 255,55 g/mol
MDL: MFCD06659920
CAS-Nummer: 850567-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR