3-Nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-2347-1G PN-2347-5G
COBBPN-2347-1GEA 0 EUR
COBBPN-2347-1G COBBPN-2347-5G
3-Nitrophenylboronsäurepinakolester
3-Nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₆BNO₄
MDL: MFCD05663879
CAS-Nummer: 68716-48-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR