3,5-Dimethoxyphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(3,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 3,5-Dimethoxyphenylboronic acid Pinacol ester
PN-2622-5G
PN-2622-1G
COBBPN-2622-5GEA
236
EUR
COBBPN-2622-5G
COBBPN-2622-1G
3,5-Dimethoxyphenylboronsäurepinakolester
3,5-Dimethoxyphenylboronsäurepinakolester
Formel:
C₁₄H₂₁BO₄ Molecular Weight: 264.13 g/mol |
MDL:
MFCD05865191 CAS-Nummer: 365564-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...