2-Hydroxyphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

M-Clarity™ Qualitätsstufe = MQ100

522554-1G 522554-5G
SIAL522554-1GEA 46.5 EUR
SIAL522554-1G SIAL522554-5G
2-Hydroxyphenylboronsäurepinakolester, Sigma-Aldrich®
2-Hydroxyphenylboronsäurepinakolester
Formel: C₁₂H₁₇BO₃
Molecular Weight: 220,08 g/mol
MDL: MFCD02093754
CAS-Nummer: 269409-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR