2-Hydroxyphenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

010835-5G FL10835.20 010835-1G
FLUO010835-5GEA 170 EUR
FLUO010835-5G CHMPFL10835.20 FLUO010835-1G
2-Hydroxyphenylboronsäurepinakolester
2-Hydroxyphenylboronsäurepinakolester
Formel: C₁₂H₁₇BO₃
Molecular Weight: 220.08 g/mol
MDL: MFCD02093754
CAS-Nummer: 269409-97-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR