3-Hydroxyphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

M-Clarity™ Qualitätsstufe = MQ100

522562-1G 522562-5G
SIAL522562-1GEA 36.4 EUR
SIAL522562-1G SIAL522562-5G
3-Hydroxyphenylboronsäurepinakolester, Sigma-Aldrich®
3-Hydroxyphenylboronsäurepinakolester
Formel: C₁₂H₁₇BO₃
Molecular Weight: 220,08 g/mol
MDL: MFCD02093755
CAS-Nummer: 214360-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR