3-Hydroxyphenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

F010836-5G FL10836.20 010836-25G 010836-1G
FLUOF010836-5GEA 137 EUR
FLUOF010836-5G CHMPFL10836.20 FLUO010836-25G FLUO010836-1G
3-Hydroxyphenylboronsäurepinakolester
3-Hydroxyphenylboronsäurepinakolester
Formel: C₁₂H₁₇BO₃
Molecular Weight: 220.08 g/mol
MDL: MFCD02093755
CAS-Nummer: 214360-76-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR