4-Brom-2-fluor-5-methoxyphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
PN-5635-1G
PN-5635-100M
COBBPN-5635-1GEA
1370
EUR
COBBPN-5635-1G
COBBPN-5635-100M
4-Brom-2-fluor-5-methoxyphenylboronsäurepinakolester
4-Brom-2-fluor-5-methoxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BBrFO₃ Molecular Weight: 330.99 g/mol |
MDL:
MFCD11855986 CAS-Nummer: 1150271-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...