2,4-Dichlorphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(2,4-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-2267-1G
PN-2267-5G
COBBPN-2267-1GEA
137
EUR
COBBPN-2267-1G
COBBPN-2267-5G
2,4-Dichlorphenylboronsäurepinakolester
2,4-Dichlorphenylboronsäurepinakolester
Formel:
C₁₂H₁₅BCl₂O₂ Molecular Weight: 272,97 g/mol |
MDL:
MFCD06795660 CAS-Nummer: 68716-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...