2-Chlorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chlorophenylboronic acid Pinacol ester, 2-(2-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

PN-2259-1G PN-2259-5G
COBBPN-2259-1GEA 50.8 EUR
COBBPN-2259-1G COBBPN-2259-5G
2-Chlorphenylboronsäurepinakolester
2-Chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₆BClO₂
Molecular Weight: 238,52 g/mol
MDL: MFCD05663846
CAS-Nummer: 870195-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR