3,4-Dichlorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3,4-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,4-Dichlorophenylboronic acid, pinacol ester

PN-2646-5G PN-2646-1G
COBBPN-2646-5GEA 324 EUR
COBBPN-2646-5G COBBPN-2646-1G
3,4-Dichlorphenylboronsäurepinakolester
3,4-Dichlorphenylboronsäurepinakolester
Formel: C₁₂H₁₅BCl₂O₂
Molecular Weight: 272,97 g/mol
MDL: MFCD05663853
CAS-Nummer: 401797-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR