3,5-Dichlorphenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
2-(3,5-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
021834-5G
021834-25G
FLUO021834-5GEA
115
EUR
FLUO021834-5G
FLUO021834-25G
3,5-Dichlorphenylboronsäurepinakolester
3,5-Dichlorphenylboronsäurepinakolester
Formel:
C₁₂H₁₅BCl₂O₂ Molecular Weight: 272.97 g/mol |
MDL:
MFCD05863906 CAS-Nummer: 68716-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...