3-Chlorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Chlorophenylboronic acid, pinacol ester

PN-2260-1G PN-2260-5G
COBBPN-2260-1GEA 0 EUR
COBBPN-2260-1G COBBPN-2260-5G
3-Chlorphenylboronsäurepinakolester
3-Chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₆BClO₂
MDL: MFCD05663877
CAS-Nummer: 635305-47-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR