4-Chlorphenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(4-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chlorophenylboronic acid Pinacol ester, 4-Chlorobenzeneboronic acid pinacol ester, 1-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
M-Clarity™ Qualitätsstufe = MQ100
632724-10G
632724-5G
SIAL632724-10GEA
168
EUR
SIAL632724-10G
SIAL632724-5G
4-Chlorphenylboronsäurepinakolester, Sigma-Aldrich®
4-Chlorphenylboronsäurepinakolester
Formel:
C₁₂H₁₆BClO₂ Molecular Weight: 238,52 g/mol |
MDL:
MFCD05663875 CAS-Nummer: 195062-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...