2-Amino-5-chlorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Amino-5-chlorophenyl boronic acid pinacol ester, 4-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

PN-8881-1G PN-8881-250M PN-8881-5G
COBBPN-8881-1GEA 449 EUR
COBBPN-8881-1G COBBPN-8881-250M COBBPN-8881-5G
2-Amino-5-chlorphenylboronsäurepinakolester
2-Amino-5-chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₂
Molecular Weight: 253,54 g/mol
MDL: MFCD06795672
CAS-Nummer: 1073371-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR