3-Amino-4-chlorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Amino-4-chlorobenzeneboronic acid pinacol ester, 3-Amino-4-chlorophenylboronic acid, pinacol ester, 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

PN-2602-5G PN-2602-1G
COBBPN-2602-5GEA 961 EUR
COBBPN-2602-5G COBBPN-2602-1G
3-Amino-4-chlorphenylboronsäurepinakolester
3-Amino-4-chlorphenylboronsäurepinakolester
Formel: C₁₂H₁₇BClNO₂
Molecular Weight: 253,54 g/mol
MDL: MFCD06659926
CAS-Nummer: 850567-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR